1-Methylnaphthalene 96%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
127161000 127165000 127160025
ACRO127161000EA 28.4 EUR
ACRO127161000 ACRO127165000 ACRO127160025
1-Methylnaphthalene 96%
1-Methylnaphthalene
Formule: C₁₁H₁₀
Kookpunt: 240…243 °C (760 mmHg)
Smeltpunt: –22 °C
Dichtheid: 1,02 g/cm³
Vlampunt: 82 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004034
CAS nummer: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.6100 to 1.6180 (20°C, 589 nm)
GC ≥99.4 %
Color scale ≤10 APHA
Aldehyde (C3H6O) ≤0.01