Tetrahydrofuraan ≥99.7% unstabilised for spectrophotometry
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Butylene oxide, THF, Diethylene oxide
Packaged under argon in resealable ChemSeal™ bottles.
41819.AK
41819.K2
41819.M1
41819.AKEA
45.1
EUR
41819.AK
41819.K2
41819.M1
Tetrahydrofuraan ≥99.7% unstabilised for spectrophotometry
Tetrahydrofuraan
Purity: 99.7+%
CAS No.: 109-99-9
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.889
Refractive Index: 1.4070
Appearance: Colourless
Odour: Ether-like
Melting Point: -108°
Boiling Point: 66°
Beilstein Reference: 102391
Merck Reference: 14,9211
UN No.: 2056
MDL No.: MFCD00005356
Formule:
C₄H₈O Kookpunt: 66 °C Smeltpunt: –108 °C Dichtheid: 0,889 Vlampunt: –17 °C (1 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00005356 CAS nummer: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3,II Bereik: 05-2114615188-48 Merck Index: 14,9211 |