Tetrahydrofuraan
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Butylene oxide, THF, Diethylene oxide
6788.1
6788.2
ROTH6788.1EA
0
EUR
ROTH6788.1
ROTH6788.2
Tetrahydrofuraan
Tetrahydrofuraan
Formule:
C₄H₈O Kookpunt: 66 °C (1013 hPa) Dichtheid: 0,888 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00005356 CAS nummer: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3,II Bereik: 05-2114615188-48 Merck Index: 12,09351 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...