2-Bromo-5-nitropyridine 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
357670010 357670050
ACRO357670010EA 28.1 EUR
ACRO357670010 ACRO357670050
2-Bromo-5-nitropyridine 98%
2-Bromo-5-nitropyridine
Formule: C₅H₃BrN₂O₂
MW: 203 g/mol
Kookpunt: 145…147 °C (10 mmHg)
Smeltpunt: 137…141 °C
MDL Nummer: MFCD00006222
CAS nummer: 4487-59-6
EINECS: 224-777-2
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Light yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 137 to 141 °C
GC ≥97.5 %