Stirring shafts, fan shaped, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C382-02 C382-14 C382-08 C382-06
BOHLC382-02EA 341 EUR
BOHLC382-02 BOHLC382-14 BOHLC382-08 BOHLC382-06
Stirring shafts, fan shaped, BOLA
Roerders Roerassen
Stirrer shafts made of PTFE-jacketed stainless steel and the fan shaped stirring unit completely made of PTFE.
  • Suitable for stirring in narrow mouth vessels or in vessels with ground joint openings
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C