2-Chloro-5-nitropyridine 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
109740250
109741000
ACRO109740250EA
98.8
EUR
ACRO109740250
ACRO109741000
2-Chloro-5-nitropyridine 99%
2-Chloro-5-nitropyridine
Formule:
C₅H₃ClN₂O₂ Smeltpunt: 106…110 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006240 CAS nummer: 4548-45-2 EINECS: 224-908-3 |
Specification Test Results
Appearance | Light yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 106 to 110 °C |
GC | ≥98.5 % |
Titrable free acid | ≤0.005 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...