2-Chloro-6-methoxy-3-nitropyridine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

RJC01057.1 RJC01057EA RJC01057.50
MAYBRJC01057.1EA 0 EUR
MAYBRJC01057.1 MAYBRJC01057EA MAYBRJC01057.50
2-Chloro-6-methoxy-3-nitropyridine
2-Chloro-6-methoxy-3-nitropyridine
Formule: C₆H₅ClN₂O₃
MDL Nummer: MFCD00130268
CAS nummer: 38533-61-8
EINECS: 253-989-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR