3-Iodopyridine 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
137440010 137440100
ACRO137440010EA 31.4 EUR
ACRO137440010 ACRO137440100
3-Iodopyridine 99%
3-Iodopyridine
Formule: C₅H₄IN
Kookpunt: 90…92 °C (14 mmHg)
Smeltpunt: 53…56 °C
Vlampunt: 106 °C
MDL Nummer: MFCD00023553
CAS nummer: 1120-90-7
EINECS: 214-322-6
Order Now
Specification Test Results
Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 56 °C
GC ≥98.5 %