2,6-Di-tert-butyl-4-methylpyridine ≥97%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 97%
CAS No.: 38222-83-2
Molecular Formula: C14H23N
Formula Weight: 205.35
Storage Temperature: 2°C to 8°C
Flash Point: 83°(181°F)
Refractive Index: 1.4765
Melting Point: 30-35°
Boiling Point: 232-234°
Beilstein Reference: 130503
MDL No.: MFCD00006305
Formule:
C₁₄H₂₃N Kookpunt: 232…234 °C Smeltpunt: 30…35 °C Vlampunt: 83 °C (181 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00006305 CAS nummer: 38222-83-2 EINECS: 253-834-4 |