2,6-Diphenylpyridine
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
BTB09880.1
BTB09880.10
BTB09880.50
MAYBBTB09880.1EA
0
EUR
MAYBBTB09880.1
MAYBBTB09880.10
MAYBBTB09880.50
2,6-Diphenylpyridine
2,6-Diphenylpyridine
Formule:
C₁₇H₁₃N MW: 231.3 g/mol Kookpunt: 185…190 °C (1 mmHg) Smeltpunt: 85 |
MDL Nummer:
MFCD00006284 CAS nummer: 3558-69-8 EINECS: 222-620-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...