2-Bromothiophene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CC13010.1 CC13010.10 CC13010.50
MAYBCC13010.1EA 0 EUR
MAYBCC13010.1 MAYBCC13010.10 MAYBCC13010.50
2-Bromothiophene
2-Bromothiophene
Formule: C₄H₃BrS
Kookpunt: ∼149 °C (1013 hPa)
Smeltpunt: –10 °C
Dichtheid: 1,71 g/cm³ (20 °C)
Vlampunt: 52 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005417
CAS nummer: 1003-09-4
EINECS: 213-699-4
UN: 2929
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR