2,5-Diiodothiophene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CD08226.1 CD08226.50
MAYBCD08226.1EA 0 EUR
MAYBCD08226.1 MAYBCD08226.50
2,5-Diiodothiophene
2,5-Diiodothiophene
Formule: C₄H₂I₂S
Kookpunt: 67…70 °C (0,05 mmHg)
Smeltpunt: 45 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00014525
CAS nummer: 625-88-7
EINECS: 210-915-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR