Thiophene 99+%, extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
138821000 138825000 138820025
ACRO138821000EA 36.8 EUR
ACRO138821000 ACRO138825000 ACRO138820025
Thiophene 99+%, extra pure
Thiophene
Formule: C₄H₄S
Kookpunt: 84 °C (760 mmHg)
Smeltpunt: –38 °C
Dichtheid: 1,065 g/cm³
Vlampunt: –9 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005413
CAS nummer: 110-02-1
EINECS: 203-729-4
UN: 2414
ADR: 3,II
Merck Index: 12,09490
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5290 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤50 APHA
Benzene ≤0.1 % (GC)