Special stirrer bearings, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C430-20 C430-28
BOHLC430-20EA 2310 EUR
BOHLC430-20 BOHLC430-28
Special stirrer bearings, BOLA
Roerders Roergereedschap
Stirrer bearings made of PTFE, provides a safe seat in the reactor lid. The stirrer shaft is held by an invisible shaft guide made of borosilicate glass which has an adjustable vacuum sealing.
  • Chemical resistance
  • Temperature resistance: −50 to 150 °C
  • Height adjustment can be done by lock nut
Suitable for long-term use. For all stirrer shafts made of stainless steel, glass or for stirrer shafts with a diameter of 8 or 10 mm.