3-Azetidinecarboxylic acid

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3-Azetanecarboxylic acid, Azetidine-3-carboxylic acid

4010451.0050 4010451.0250 4010451.1000
BACE4010451.0050EA 0 EUR
BACE4010451.0050 BACE4010451.0250 BACE4010451.1000
3-Azetidinecarboxylic acid
3-Azetidinecarboxylic acid
Formule: C₄H₇NO₂
MDL Nummer: MFCD00191763
CAS nummer: 36476-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR