Lithiumaluminiumhydride 2.4 mol/l in THF, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Aluminium lithium hydride, Lithium tetrahydridoaluminate
377321000 377328000
377321000.EA 49.2 EUR
377321000. 377328000.
Lithiumaluminiumhydride 2.4 mol/l in THF, AcroSeal®
Lithiumaluminiumhydride
Formule: H₄AlLi
MW: 37.95 g/mol
Smeltpunt: 125
Vlampunt: –17 °C
MDL Nummer: MFCD00011075
CAS nummer: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3 I
Merck Index: 13,00348
Order Now
Specification Test Results
Appearance Colorless to grey Solution (May develop some turbidity or precipitate)
Strength 9.5 to 10.5 % (as LiAlH4)