Scrapers for industry
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8300-4002 8300-4001
233-0194EA 9.1 EUR
233-0194 BURK8300-4001
Scrapers for industry
Schrapers
These scrapers made of green PP are ideal for industrial applications, such as pharmaceuticals, biotechnology, foods, chemistry and other industries.
  • Autoclavable up to 121 °C for max. 20 min.
  • With fixed handle
  • Anti-static