Scrapers for foodstuffs
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BURK8300-4011EA 7 EUR
BURK8300-4011 BURK8300-4010
Scrapers for foodstuffs
Schrapers
These scrapers made of white PP are ideal for use with foodstuffs.
  • Autoclavable up to 121 °C for max. 20 min.
  • With fixed handle