Filling scoops for foodstuff
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8300-5010 8300-5012 8300-5011
BURK8300-5010EA 7.8 EUR
BURK8300-5010 BURK8300-5012 233-0195
Filling scoops for foodstuff
Scheppen
PP, white. These durable scoops are ideal for use in food processing, agriculture, fish farming, sampling and veterinary use.
  • Easy to clean
  • Smooth, non stick surface
  • Autoclavable at 121 °C