Safety lancets, BD Microtainer® QuikHeel™
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
368103 368102
BDAM368103EA 475 EUR
BDAM368103 BDAM368102
Safety lancets, BD Microtainer® QuikHeel™
Bloedlancetten
This lancet is used for capillary blood collection from the heels of premature babies, newborns and infants.
  • Extra thin steel blade with a defined incision depth and width
  • Fine, clean, surgical incision at the touch of a button for a proper blood flow
  • Concealed blade remains enclosed by the housing and eliminates any possibility of injury or reuse
  • Tapered grips for a better localisation of the puncture site
Verpakking: Individually wrapped in sterile packaging.