Burette and condenser clamps, with shaft
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5628 5629
BOCH5628EA 32 EUR
BOCH5628 BOCH5629
Burette and condenser clamps, with shaft
Klemmen Klemmen voor algemeen gebruik
Burette clamps made of chromium plated steel.
  • Fingers are coated with PVC to protect clamped items
  • With shaft