On-column syringes
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
000376 000380
SGEA000376EA 145 EUR
SGEA000376 SGEA000380
On-column syringes
Spuiten Injectiespuiten voor chromatografie
On-column syringes are suitable for general purpose applications.
When selecting an on-column syringe, the needle outside diameter (OD) must be smaller than the inside diameter (ID) of the capillary column it will be used with.