VWR® poederpakketten voor de bepaling van mangaan, reagentia voor fotometers
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85608.620EA
45.1
EUR
85608.620
85609.620
VWR® poederpakketten voor de bepaling van mangaan, reagentia voor fotometers
Milieutestkits
VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminium foil packs, are unaffected by ambient conditions and contain pre-measured powder reagents for accuracy, convenience and great solubility.
- Accurate
- Fast and convenient by easier dissolution than tablets and long shelf life
- No calibration necessary
- Beveled edge prevents reagent from getting stuck in corners
RS = Powder Pack reagent set.