Holders and adapters for BD Vacutainer® products
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
368872 364815 364815UK
BDAM368872EA 66.4 EUR
BDAM368872 BDAM364815 BDAM364815UK
Holders and adapters for BD Vacutainer® products
Houders voor Bloedafnamebuisjes
The BD Vacutainer® holders are compatible with all BD Vacutainer® blood collection system; eclipse blood collection needle, Safety-Lok™ blood collection set, push button blood collection set and multiple sample Luer adapter.
The BD Vacutainer® Luer-Lok™ access device is designed for sterile, secure and safer specimen sampling. This device provides the security of a threaded, locking Luer connection. It is also compatible with a female Luer connection or needleless IV site designed for Luer-lock access, and Luer locking Foley catheter sampling ports.