GL tube fittings, PTFE EX
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D856-18 D857-14 D856-25 D859-25 D859-18 D859-14 D858-25 D858-18 D858-14 D857-25 D856-14 D857-18
BOHLD856-18EA 49.8 EUR
BOHLD856-18 BOHLD857-14 BOHLD856-25 BOHLD859-25 BOHLD859-18 BOHLD859-14 BOHLD858-25 BOHLD858-18 BOHLD858-14 BOHLD857-25 BOHLD856-14 BOHLD857-18
GL tube fittings, PTFE EX
Fittingen en connectoren
Connector for tubing, made of PTFE-EX, with two GL screw connections. To distribute liquids or gases. Can be used as a reducer to connect different tubes or hoses.
  • Very good chemical resistance
  • Temperature resistant from −200 to +250 °C
  • Pressure resistant up to a maximum of 10 bar, vacuum suitable