Metal adapters, glass flange, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D720-10 D720-25 D720-15
BOHLD720-10EA 43.5 EUR
BOHLD720-10 BOHLD720-25 BOHLD720-15
Metal adapters, glass flange, BOLA
Afdichtingen Pakkingen
PTFE/Silicone.
  • Temperature resistance: from −60 to 230 °C
  • Universal chemical resistance
  • Pressure: low
  • Vacuum: suitable
Made of PTFE and silicone, circular PTFE collar facilitates assembly and assures exact placement of the gasket. The elasticity for expansions and contractions is given by a special kind of silicone which is placed behind PTFE sealing lips. Universal chemical resistance since the product is only exposed to PTFE.
For HWS® 'Adaptor, metal, for flexible metallic hose' for a reliable sealing between glass flange and metal adaptor. Ideal for sealing temper connections, e.g. on double walled vessels.