Bottles, wide neck, safety coated, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216627 216629 216637 216639
WHEA216627EA 266 EUR
WHEA216627 WHEA216629 WHEA216637 WHEA216639
Bottles, wide neck, safety coated, WHEATON®
Flessen Glazen flessen
Bottles, made from clear soda-lime glass that conforms to USP Type III requirements. Wide neck for use with solid sample.
  • Plastisol coating for safety
  • Non-slip coating allows for greater ease and stability when handling wet or dry
  • Black phenolic poly-vinyl lined cap
Bottles with screw caps come attached to help maintain cleanliness.