Tubings, PFA, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S1811-02 S1811-04 S1811-06 S1811-28 S1811-18 S1811-40 S1811-24 S1811-30 S1811-10 S1811-14 S1811-50 S1811-12 S1811-08 S1811-26 S1811-22 S1811-20 S1811-16 S1811-05 S1811-07 S1811-03
BOHLS1811-02EA 15.2 EUR
BOHLS1811-02 BOHLS1811-04 228-1197 BOHLS1811-28 BOHLS1811-18 BOHLS1811-40 BOHLS1811-24 BOHLS1811-30 BOHLS1811-10 BOHLS1811-14 BOHLS1811-50 BOHLS1811-12 BOHLS1811-08 BOHLS1811-26 BOHLS1811-22 BOHLS1811-20 BOHLS1811-16 BOHLS1811-05 BOHLS1811-07 BOHLS1811-03
Tubings, PFA, BOLA
Slangen Transfer Tubing
Made of PFA, transparent
  • Temperature resistance: −200 to +260°C
  • Gastighttubing
  • Almost universal chemical resistance
  • Resistant to irradiation and weather
  • Sterilized in autoclaves
  • Mechanical strength even at high temperatures
Applications include perfect tubing for aggressive and pure liquids or gases. Good bending radius and burst pressure.