Reagent bottles, with screw cap, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
220163 220223 220283
WHEA220163EA 120 EUR
WHEA220163 WHEA220223 WHEA220283
Reagent bottles, with screw cap, WHEATON®
Flessen Glazen flessen
Manufactured from USP Type I borosilicate glass, with large writing patch.
  • Features no-drip pour lip
  • Pre-attached polypropylene screw cap with PTFE liner
  • Autoclavable