Tubing, FEP, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S1815-04 S1815-40 S1815-56 S1815-08 S1815-20 S1815-48 S1815-36 S1815-60 S1815-12 S1815-28 S1815-68 S1815-44 S1815-32 S1815-24 S1815-16 S1815-07
BOHLS1815-04EA 6 EUR
BOHLS1815-04 BOHLS1815-40 BOHLS1815-56 BOHLS1815-08 BOHLS1815-20 BOHLS1815-48 BOHLS1815-36 BOHLS1815-60 BOHLS1815-12 BOHLS1815-28 BOHLS1815-68 BOHLS1815-44 BOHLS1815-32 BOHLS1815-24 BOHLS1815-16 BOHLS1815-07
Tubing, FEP, BOLA
Slangen Transfer Tubing
PFA, transparent.
  • Gastight tubing
  • Almost universal chemical resistance
  • Good sliding and dielectric characteristics
  • Can be sterilized in autoclaves
  • Resistant to irradiation and weather
Applications include perfect tubing for aggressive and pure liquids or gases. It has good bending radius and burst pressure.
Temperature resistance: −200 to +260 °C.