Capillary Tubing, PEEK, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S1817-16 S1817-20 S1817-08 S1817-12
BOHLS1817-16EA 52.2 EUR
BOHLS1817-16 BOHLS1817-20 BOHLS1817-08 BOHLS1817-12
Capillary Tubing, PEEK, BOLA
Slangen Chromatography Tubing
PEEK.
  • Temperature resistance: −50 to +250 °C
  • Universal chemical resistance
  • Metal-free
  • Corrosion-proof
  • High pressure resistance
  • Biocompatible
  • High temperature resistance (melting point +334 °C)
Flexible, brown high-pressure tubing for almost all organic or inorganic liquids.