Bottles, narrow neck, round, HDPE, with screw caps, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
221153 221154 221163 221164
WHEA221153EA 170 EUR
WHEA221153 WHEA221154 WHEA221163 WHEA221164
Bottles, narrow neck, round, HDPE, with screw caps, WHEATON®
Flessen Kunststof flessen
HDPE, natural or white.
  • Good chemical resistance
  • Narrow mouth ideal for liquids
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner
Order Now
Specificaties
Environmentally Preferable