Snap caps for sample bottles, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
242612 242616 242619 242615
WHEA242612EA 69.9 EUR
WHEA242612 WHEA242616 WHEA242619 WHEA242615
Snap caps for sample bottles, WHEATON®
Sluitingen Flesdoppen
Natural low density polyethylene.
  • All sizes have tabs for easy removal
  • Good chemical resistance
  • Good temperature resistance
For WHEATON snap cap sample bottles.