Surgical scalpels and blades, Sabre
Leverancier: SWANN MORTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0570 0565 0563
SWAN0570EA 18.4 EUR
SWAN0570 SWAN0565 233-0163
Surgical scalpels and blades, Sabre
Scalpels Scalpels
Carbon steel blades. These blades are available separately without handles or as ready-to-use scalpels with PS handle. They are ideal for use during Podiatry and Chiropody treatments.
  • Scalpels are sterile and diposable
  • PS handles carry a graduated measure (for indication only) on the reverse side
  • Closed edge blades are ideal for 'paring' or 'scraping' skin within Podiatry and Chiropody treatments
Verpakking: Sterile scalpels are individually packaged into a transparent thermo formed tray providing improved product visibility.
Blades only are available in individual foil packs (sterile) or in foil packets of five (non sterile).