Spouts for jerricans
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
610499 610500 610501 610502 610503 610504
590-0845EA 17.2 EUR
590-0845 590-0846 590-0847 590-0848 590-0849 SCTV610504
Spouts for jerricans
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
For simple and safe pouring of fluids from containers.
  • Plastic
  • Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S55, S60/61, and S40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.