Syringes
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008787 008987
SGEA008787EA 155 EUR
SGEA008787 SGEA008987
Syringes
Spuiten Injectiespuiten voor chromatografie
These syringes are made from borosilicate glass. They are used as a replacement for pump syringes.
  • Accuracy and reproducibility of ±1%
  • Scale length: 60 mm
Thread in plunger button: 6–32 UNC.