Closures for diagnostic bottles, I-Loc™, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240676-01 240676-04 240676-05 240676-02
WHEA240676-01EA 121 EUR
WHEA240676-01 WHEA240676-04 WHEA240676-05 WHEA240676-02
Closures for diagnostic bottles, I-Loc™, WHEATON®
Sluitingen Flesdoppen
Closure made of PP and available with bromobutyl/50 stoppers.
  • Ideal for use with diagnostic bottles
  • Eliminates use of separate aluminum seal and rubber stopper