Tubings, PFA, corrugated, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S1820-06 S1820-02 S1820-04 S1820-23 S1820-16 S1820-14 S1820-01 S1820-10 S1820-08
BOHLS1820-06EA 78.4 EUR
BOHLS1820-06 BOHLS1820-02 BOHLS1820-04 BOHLS1820-23 BOHLS1820-16 BOHLS1820-14 BOHLS1820-01 BOHLS1820-10 BOHLS1820-08
Tubings, PFA, corrugated, BOLA
Slangen Transfer Tubing
Made of PFA, transparent.
  • Tight bending radius only causes little cross-section reduction
  • Non porous and translucent
  • Almost universal chemical resistance
  • Highly flexible
These tubing contains circular corrugations around the longitudinal axis. It can be shortened easily by means of a tubing cutter. Applications include perfect tubing for aggressive and pure liquids or gases and good bending radius.