Stoppers for BOD bottles, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
227670 227672
WHEA227670EA 129 EUR
WHEA227670 WHEA227672
Stoppers for BOD bottles, WHEATON®
Stoppen Glazen stoppen
Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For 60 ml and 300 ml BOD bottles.