Tweezers, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H909-04 H912-02 H912-04 H912-06 H909-06 H909-02
BOHLH909-04EA 28.8 EUR
BOHLH909-04 BOHLH912-02 BOHLH912-04 BOHLH912-06 BOHLH909-06 BOHLH909-02
Tweezers, BOLA
Pincetten
Precast tweezers made of PTFE.
  • With pointed or blunt ends
  • Universal chemical resistance
  • Temperature resistant from −200 to +250 °C