Double spatulas, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H915-06 H915-02 H915-04
BOHLH915-06EA 25.2 EUR
BOHLH915-06 BOHLH915-02 BOHLH915-04
Double spatulas, BOLA
Spatels
PTFE, white.
  • With tapered ends
  • Universal chemical resistance
  • Temperature resistant from −200 to +250 °C
  • Suitable for food contact (FDA conform)