Trypsinising flasks, with screw caps, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
355394 355395 355397 355392 355445 355442 355444
WHEA355394EA 129 EUR
WHEA355394 WHEA355395 WHEA355397 WHEA355392 WHEA355445 WHEA355442 WHEA355444
Trypsinising flasks, with screw caps, WHEATON®
Kolven Trypsinisatie kolven
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
Certificaten: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.