Scrapers, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H916-08 H916-06 H916-02
BOHLH916-08EA 415 EUR
BOHLH916-08 BOHLH916-06 BOHLH916-02
Scrapers, BOLA
Schrapers
Scrapers made of PTFE with tapered end.
  • Universal chemical resistance
  • Temperature resistant from −200 to+250 °C
  • Ideal handling due to big handle and wide blade
  • For a very gentle peeling of products
Handle diameter: 20 mm