Zuig/drukballonnen
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3910 3911 3912 3913
KART3910EA 38.1 EUR
KART3910 KART3911 KART3912 KART3913
Zuig/drukballonnen
Pipetten Pipetvullerballonnen
Gemaakt van rubber, rood.
Gebruikt om b.v. automatische buretten te vullen.
Double balls with net are suitable for automatic burettes and flash chromatography columns.
Tube length: 33,5 cm