Caps for bottles, open top, Microlink®, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240106 240235-01 240250 240241 240241-01 240250-01 240251-01 240256-01 240112 240114 240200
WHEA240106EA 128 EUR
WHEA240106 WHEA240235-01 WHEA240250 WHEA240241 WHEA240241-01 WHEA240250-01 WHEA240251-01 WHEA240256-01 WHEA240112 WHEA240114 WHEA240200
Caps for bottles, open top, Microlink®, WHEATON®
Sluitingen Flesdoppen
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
  • PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
  • Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.