Stopcocks, miniature
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOLA
BOHLF731-10EA 211 EUR
BOHLF731-10 BOHLF731-12 BOHLF731-02 BOHLF730-06 BOHLF731-08 BOHLF730-08 BOHLF730-02 BOHLF731-14 BOHLF731-04 BOHLF730-04 BOHLF731-16 BOHLF731-06
Stopcocks, miniature
Afsluitkranen
Miniature stopcocks of PTFE with male or female connection threads and stopcock plug of FEP.
  • Temperature resistance from -200 to 205°C
  • Universally chemical resistant
  • Pressure upto 8 bar