Cleanroom gelaatsmaskers, BioClean MEA™, PP/PE
Leverancier: Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MEA210-0 MEA210-1
113-1488EA 1180 EUR
113-1488 113-8160
Cleanroom gelaatsmaskers, BioClean MEA™, PP/PE
Maskers Cleanroom Maskers
Niet-steriele of steriele gelaatsmaskers.
  • Ultralage deeltjesafgifte
  • PU-oorlussen
  • Clip voor sluiting aan achterkant hoofd
  • Volledig gesloten, vervormbaar neusstuk
Geschikt voor cleanrooms ISO Klasse 4.
Certificaten: Manufactured in a facility holding ISO 9001:2015. Processed in an NEBB certified ISO class 4 cleanroom environment. Sampling and inspection in accordance with ISO 2859-3:1994. Tested against standard ASTM F2101 for bacterial filtration efficiency (BFE).
Verpakking: Non sterile masks are bulk packed. Sterile masks are individually packaged.