Micro-fluidic platform, SilFlow™
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
123722 123725 123730 123731 123732 123733 123734 123735 1237031 123704 123705 123706 123707 123709 123717 123720 123721 123723 123724 123750 123751 123752 123753 123755 123708 123713 123715 123742 123743 123744
548-1780EA 766 EUR
548-1780 548-1783 548-1785 548-1786 548-1787 548-1788 548-1789 548-1790 548-1769 548-1770 548-1771 548-1772 548-1773 548-1774 548-1777 548-1778 548-1779 548-1781 548-1782 SGEA123750 SGEA123751 SGEA123752 SGEA123753 SGEA123755 548-1799 548-1800 548-1801 548-1802 SGEA123743 SGEA123744
Micro-fluidic platform, SilFlow™
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
SilFlow™ microchannel device (MCD) options for three-port GC splitters, four-port GC splitters and a Deans' switch MCD, perfect for multi-dimensional analyses. SilFlow™ can be incorporated into the system without impacting the chromatography.
  • Chemically-deactivated stainless steel channels avoiding active sites experienced with conventional connections
  • Conventional graphite or Vespel® ferrules cannot be used with the SilFlow™ MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
  • SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection, giving optimised peak shapes
  • Can be operated at pressures greater than 25000 psi
  • SilFlow™ tracks oven temperature up to 20 °C/min. The design of SilFlow™ alleviates cold spots and sample condensation
Bestelinformatie: SilFlow™ kits contain: SilFlow™ microchannel device (MCD), SilTite™ FingerTite tool, mounting bracket, appropriately sized ferrules, nuts and blanking ferrules for set up. 1,1 mm Ø tubing is not supplied in the splitter kits and must be ordered separately.