Jars, wide mouth, with screw cap, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F109150000 F109160000 F109130000 F109140000 F109170000
BELAF109150000EA 146 EUR
BELAF109150000 BELAF109160000 BELAF109130000 BELAF109140000 BELAF109170000
Jars, wide mouth, with screw cap, SP Bel-Art
Potjes Potjes voor algemeen gebruik
PP
  • High shock resistance
  • Screw caps with unique grip lugs
  • With G-threaded neck and removable leakproof silicone rubber ring seal
These mason jars are are ideal for industrial waste sampling, laboratory uses and long term storage.