Gas inlet tubes, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N1502-02 N1502-04 N1502-06
BOHLN1502-02EA 26.2 EUR
BOHLN1502-02 BOHLN1502-04 BOHLN1502-06
Gas inlet tubes, BOLA
Buisjes Gasinlaatbuizen
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
For constructing a gas washing equipment. Tube with Int.Ø 5 mm, one side with hose connector dia. 9 mm for connecting tubing, other side with thread M 8×1 for connecting a gas frit or a gas distributor.